Learn More
Water for Injection, Waternation
Product Code. 17888020
Click to view available options
Quantity:
10 x 100 mL
10 x 500 mL
6 x 1000 mL
Sterile Water for Injection, particle free. Complies with the requirements of ASTMD1193-06.
  • Ready to use
  • No chemical additives
  • Batch traceability
  • Certificate of conformity and analysis
  • Reduce expensive test fails due to impurities
  • High performance
  • High accuracy
  • Suppress water-forming side reactions
  • Provide consistent performance across many sample types
The MLG range, a comprehensive suite of five high grade pure waters, in assorted sizes, ranging from simple deionised to molecular biology grades, catering to the majority of pure water lab uses.
Chemical Identifiers
CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O
Specifications
pH 7
Conductivity 0.1 μS/cm
Grade Particle Free
Packaging PET Plastic Bottle
Sterility Sterile
Certificate of Analysis Yes
Quantity 10 x 100 mL
For Use With (Application) Water for Irrigation, Cell Culture