Learn More
BUCHI Water Jet Pump For Hydrolysis Unit B-411 E-416
Brand: BUCHI 002913
Additional Details : Weight : 0.06000kg
Specifications
Water Jet Pump | |
Used for aspiration of harmful vapors from the digestion unit | |
For Hydrolysis Unit B-411, E-416 | |
Plastic |