Learn More
BINDER™ Water Supply Set
Water Supply Set consisting of fresh- and waste-water containers (20 liters each), cabling and pump.
Brand: BINDER™ 8012-1846
13501.72 SEK valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 10.00000kg
Description
- External
- Includes fresh and waste water containers (20 L), cabling, and pump
Specifications
Water supply set - external, free-standing,consisting of fresh- and waste-water containers (20 liters each), cabling and pump | |
Plastic | |
Fresh- and Waste Water Containers (20 Liters Each), Cabling and Pump |
BINDER Chambers Series KBWF, KBF, KBF P, KBF LQC, MKF 56 | |
Water Supply |