Learn More
WEE2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Bio-Techne NBP3-17636-100UL
 View more versions of this product
Product Code. 18314536
  • 5775.00 SEK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
WEE2 Polyclonal antibody specifically detects WEE2 in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
WEE2
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
EC 2.7.10, EC 2.7.10.2, FLJ16107, WEE1 homolog 2 (S. pombe), WEE1B, Wee1B kinase, Wee1-like protein kinase 1B, wee1-like protein kinase 2
This antibody was developed against Recombinant Protein corresponding to amino acids: DIDKELRQKLNFSYCEETEIEGQKKVEESREASSQTPEKGEVQDSEAKGTPPWTPLSNVHELDTSSEKDKESPDQILRTPVSHPLKC
100 μg
Primary
Human
Purified
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol
Rabbit
Affinity purified
RUO
494551
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.