missing translation for 'onlineSavingsMsg'
Learn More

WFDC1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

1680.00 SEK - 3415.00 SEK

Specifications

Antigen WFDC1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18321825
View Documents
Bio-Techne
NBP3-15532-20UL
20 μg
1680.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18377786
View Documents
Bio-Techne
NBP3-15532-100UL
100 μg
3415.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

WFDC1 Polyclonal antibody specifically detects WFDC1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

WFDC1
Polyclonal
Purified
RUO
Human, Mouse
ps20 growth inhibitor, PS20Prostate stromal protein ps20, WAP four-disulfide core domain 1, WAP four-disulfide core domain 1 homolog, WAP four-disulfide core domain protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 111-220 of human WFDC1 (NP_067020.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Cell Biology
PBS, 50% glycerol, pH7.3
58189
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.