Learn More
Cytiva Whatman™ Type WCN Cellulose Nitrate Membranes
Recommended for majority of routine applications, this membrane is manufactured under strictly controlled conditions.  Whatman™ Type WCN Cellulose Nitrate Membranes narrow pore size distribution for improved surface capture and analysis.
1145.00 SEK - 2845.00 SEK
Specifications
Autoclavable Autoclavable
Color White
Material Cellulose Nitrate
View More Specs
Products 26
Product Code Brand Diameter (Metric) Pore Size Quantity Price Quantity & Availability  
Product Code Brand Diameter (Metric) Pore Size Quantity Price Quantity & Availability  
11384854
missing translation for 'viewDocuments' Promotion Details
Cytiva
7190-002
25 mm 1.0 μm 100/Pk.
1515.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11324864
missing translation for 'viewDocuments' Promotion Details
Cytiva
7193-002
25 mm 3.0 μm 100/Pk.
1520.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
10154211
missing translation for 'viewDocuments' Promotion Details
Cytiva
7184-002
25 mm 0.45 μm 100/Pk.
1205.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11354854
missing translation for 'viewDocuments' Promotion Details
Cytiva
7188-002
25 mm 0.8 μm 100/Pk.
1480.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11374834
missing translation for 'viewDocuments' Promotion Details
Cytiva
7181-004
47 mm 0.1 μm 100/Pk.
2225.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11394854
missing translation for 'viewDocuments' Promotion Details
Cytiva
7190-004
47 mm 1.0 μm 100/Pk.
2075.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11364844
missing translation for 'viewDocuments' Promotion Details
Cytiva
7184-004
47 mm 0.45 μm 100/Pk.
2260.08 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11364864
missing translation for 'viewDocuments' Promotion Details
Cytiva
7195-009
90 mm 5 μm 25/Pk.
1885.00 SEK
Pack of 25
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11314864
missing translation for 'viewDocuments' Promotion Details
Cytiva
7191-014
142 mm 1.2 μm 25/Pk.
2670.00 SEK
Pack of 25
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
10565361
missing translation for 'viewDocuments' Promotion Details
Cytiva
7188-004
47 mm 0.8 μm 100/Pk.
1965.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
10470713
missing translation for 'viewDocuments' Promotion Details
Cytiva
7195-004
47 mm 5 μm 100/Pk.
2085.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11304854
missing translation for 'viewDocuments' Promotion Details
Cytiva
7184-014
142 mm 0.45 μm 25/Pk.
2685.00 SEK
Pack of 25
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
10652972
missing translation for 'viewDocuments' Promotion Details
Cytiva
7193-004
47 mm 3.0 μm 100/Pk.
2150.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11334854
missing translation for 'viewDocuments' Promotion Details
Cytiva
7186-004
47 mm 0.65 μm 100/Pk.
2125.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
11364854
missing translation for 'viewDocuments' Promotion Details
Cytiva
7188-003
37 mm 0.8 μm 100/Pk.
1960.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11384834
missing translation for 'viewDocuments' Promotion Details
Cytiva
7182-001
13 mm 0.2 μm 100/Pk.
1360.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11364834
missing translation for 'viewDocuments' Promotion Details
Cytiva
7181-002
25 mm 0.1 μm 100/Pk.
1600.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
10777222
missing translation for 'viewDocuments' Promotion Details
Cytiva
7182-004
47 mm 0.2 μm 100/Pk.
2385.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11334844
missing translation for 'viewDocuments' Promotion Details
Cytiva
7184-001
13 mm 0.45 μm 100/Pk.
1145.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11344864
missing translation for 'viewDocuments' Promotion Details
Cytiva
7195-002
25 mm 5 μm 100/Pk.
1375.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11394844
missing translation for 'viewDocuments' Promotion Details
Cytiva
7184-009
90 mm 0.45 μm 25/Pk.
2025.00 SEK
Pack of 25
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11324844
missing translation for 'viewDocuments' Promotion Details
Cytiva
7182-014
142 mm 0.2 μm 25/Pk.
2845.00 SEK
Pack of 25
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11395044
missing translation for 'viewDocuments' Promotion Details
Cytiva
7188-009
90 mm 0.8 μm 25/Pk.
2095.00 SEK
Pack of 25
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
10694112
missing translation for 'viewDocuments' Promotion Details
Cytiva
7182-002
25 mm 0.2 μm 100/Pk.
1500.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11314844
missing translation for 'viewDocuments' Promotion Details
Cytiva
7182-009
90 mm 0.2 μm 25/Pk.
1560.00 SEK
Pack of 25
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
11374844
missing translation for 'viewDocuments' Promotion Details
Cytiva
7184-005
50 mm 0.45 μm 100/Pk.
2015.00 SEK
Pack of 100
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
Description
Description
Recommended for majority of routine applications, this membrane is manufactured under strictly controlled conditions. Whatman™ Type WCN Cellulose Nitrate Membranes narrow pore size distribution for improved surface capture and analysis.
  • Higher strength and flexibility - Most membranes are inherently brittle and difficult to handle; it is not uncommon for filters to be damaged during loading into holders or while in use. Whatman cellulose nitrate membrane filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity. These membranes are among the strongest of their type available, as measured and compared by burst pressure tests.
  • Low extractable levels - The level of extractables in membrane filters has become more important with advances in filtration or adsorption techniques. In particular, pharmaceutical, immunological, and biomedical tissue culture and trace analysis applications can be adversely affected by high extractable levels. Whatman cellulose nitrate membrane filters have a low level of extractables, generally below that of other membranes of a similar type.
  • Narrow pore size distribution - One of the major features of Whatman membrane filters is the narrow distribution of pore sizes. The rated pore size of these membranes is closely controlled due to the advanced manufacturing and control system. Additionally, the batch-to-batch variation is minimized providing more consistent laboratory results.
  • Increased temperature stability - Membrane filters are normally autoclaved at 121°C without loss of integrity. Cellulose nitrate membranes are supplied as circles, sheets, or reels.
  • Reduced shrinkage - Excessive shrinkage can cause problems during autoclaving and is often the cause of membranes tearing in their holders after autoclaving. It may also cause a reduction in flow rate and total throughput. Whatman membranes exhibit a low shrinkage during autoclaving.
  • White plain filters - This is the standard membrane filter for the majority of laboratory applications involving particles and cells in the range of 0.1 μm to 12.0 μm. The residue after filtration is found to be almost completely on the surface of the membrane and allows physical recovery of deposits and microscopic examination.
Specifications
Specifications
Autoclavable
White
Cellulose Nitrate