missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Vacuum-Type Glass Membrane Holder

Produced from borosilicate glass and available with a choice of support screen. Whatman™ Vacuum-Type Glass Membrane Holder is suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.

Brand:  Cytiva 1960-002

Additional Details : Weight : 0.16000kg

 View more versions of this product

Product Code. 11300644

  • 4495.00 SEK / Each
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

Produced from borosilicate glass and available with a choice of support screen. Whatman™ Vacuum-Type Glass Membrane Holder is suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.

  • Made from borosilicate glass
  • Ensures secure clamping and integral sealing at membrane edge
  • Gravimetric analysis filtration
  • Ultraclean filtration
  • Bacteriological control of food, drink, pharmaceuticals, cosmetics
  • Removal of particulates in solution
  • Nucleic acid and protein analysis
  • HPLC solvent filtration
  • Specifications

    Specifications

    Recommended for filtration and biological analysis
    25 mm
    25 mL
    Borosilicate Glass
    Prefilter Membrane: 16 mm dia.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers