missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Headspace Vials Convenience Kits with Pressure Release

Newly designed top provides a tighter seal with PTFE-silicone septa

Brand:  DWK Life Sciences W225280-01

 View more versions of this product

Product Code. 12484981

  • 2625.00 SEK / Pack of 100
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from Type I borosilicate glass
  • Rounded shoulders and bottoms allow for even heating and safer operation at higher pressures

Order Info

Available in convenience packs of 100. Dia. x H (10mL vials): 23x 46mm. Dia. x H (20mL vials): 23 x 75mm.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Borosilicate Glass
2.95 in.
75 mm
20 mm
23 mm
6.0 mL
Crimp Cap
100 Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.