missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Phenolic Screw Caps for Media Bottles

Product Code. 10260323
Click to view available options
Neck Finish:
38-430
This item is not returnable. View return policy
This item is not returnable. View return policy

PTFE faced (14B) styrene-butadiene rubber liner provides totally inert inner seal and surface facing the sample or product

  • Designed for the ultimate in product safety
  • Screw cap
  • Phenolic
  • Black
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Neck Finish 38-430
Certifications/Compliance ISO 9001
Autoclavable Autoclavable
For Use With (Equipment) Media bottle
Closure Liner PTFE/Rubber
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'