Learn More
DWK Life Sciences Wheaton™ Flat Bottom Flasks For Soxhlet Extraction Apparatus
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
525.00 SEK - 615.00 SEK
Specifications
Material Borosilicate Glass
For Use With (Application) For continuous extraction of analytes from a solid into an organic solvent
Standard Taper Joint 24/40
Bottom Shape Flat
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
12447111
View Documents
DWK Life Sciences
428126
125 mL
560.00 SEK
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
12457111
View Documents
DWK Life Sciences
428146
250 mL
525.00 SEK
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
12467111
View Documents
DWK Life Sciences
428164
500 mL
615.00 SEK
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
Description
Description
  • Flat bottom flask
Specifications
Specifications
Borosilicate Glass
For continuous extraction of analytes from a solid into an organic solvent
24/40
Flat