Learn More
DWK Life Sciences Wheaton™ No. 8 Silicone Stopper
Brand: DWK Life Sciences 419356
This item is not returnable.
View return policy
Description
Specifications
Designed for connecting Wheaton 47mm vacuum filter holer assemblies to glass filtering flasks | |
1 | |
Stopper (1 hole) |
8 | |
Silicone | |
8 |