Learn More
DWK Life Sciences Wheaton™ No. 8 Silicone Stopper
Brand:  DWK Life Sciences 419356
Product Code. 12401581
  • 1315.00 SEK / Pack of 6
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Designed for connecting Wheaton 47mm vacuum filter holer assemblies to glass filtering flasks
1
Stopper (1 hole)
8
Silicone
8