Learn More
DWK Life Sciences Wheaton™ Screw Thread Headspace Closures
Screw thread finish eliminates the need for crimping tools
1205.00 SEK
Specifications
Type Screw Cap
View More Specs
Products 1
Product Code Brand Septum Price Quantity & Availability  
Product Code Brand Septum Price Quantity & Availability  
15384695
View Documents
DWK Life Sciences
W224218
White PTFE/Transparent Blue Silicone
1205.00 SEK
Pack of 100
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
Description
Description
  • For use with CTC Analytics Systems requiring magnetic closures
  • Also compatible with modern Agilent, Perkin Elmer and Tekmar autosamplers that do not require transports
Specifications
Specifications
Screw Cap