missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ White Plastic Bio-Hazard Trays

Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.

580.00 SEK - 1110.00 SEK

Specifications

Color White
Material Plastic
For Use With Safe containment of spillages, lab bench protection
Includes Liner
No. per Pack 1 Each
View More Specs

Products 5
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
12387207
View Documents Promotion Details
Fisherbrand™
CSR-TO3
57 x 54 cm
790.00 SEK
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
12397207
View Documents Promotion Details
Fisherbrand™
CSR-TO4
68 x 54 cm
949.00 SEK
Each
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
 
12317217
View accessory products View Documents Promotion Details
Fisherbrand™
CSR-TO6
113 x 54 cm
1110.00 SEK
Each
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
 
12377207
View accessory products View Documents Promotion Details
Fisherbrand™
CSR-TO2
54 x 34 cm
580.00 SEK
Each
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
 
12307217
View Documents Promotion Details
Fisherbrand™
CSR-TO5
70 x 46 cm
949.00 SEK
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
Description

Description

  • Available in general purpose, biohazard and radiation safety formats
  • Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
  • All trays supplied with one free liner when purchased
  • Additional environmentally friendly APET liners are provided in packs of 25
Specifications

Specifications

White
Safe containment of spillages, lab bench protection
1 Each
Plastic
Liner
Videos
SDS
Documents

Documents

Product Certifications