Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Color | Natural |
Quantity | 1700 |
Closure Color | White |
Product Type | Unlined Cap |
Closure Liner | Unlined |
Closure Material | Polypropylene |
Closure Size | 48-400 mm |