Learn More
Kautex™ PVC Wide Neck Square Container, Brown
Fill and clean these wide neck PVC containers with ease due to the roomy neck configuration. Kautex™ PVC Wide Neck Square Container, Brown are ideal for light sensitive media.
Brand:  Kautex™ 2000084599
 View more versions of this product
Product Code. 10336031
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Member of Series 310 container family
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500 mL
54 mm
PVC
126/Cs.
Square
Wide Neck Container
Brown
Packaging, Dispensing, Sending Samples, Storage, Mailing
126
No
118.5 mm
Closure
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products