Description
- Designed for maximum comfort
- Class 1: accommodates long term use
- Durable, with a long shelf life
- Manufactured using quality materials
- Soft nose bridge, to accommodate extreme weather
- Sleek, wrap around design for wider field of view
- Suits laboratories, medical field use, maintenance and general industrial use
Specifications
Specifications
Certifications/Compliance | CE certified according to EN166: 2001 |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Quantity | Each |
UV Protection | UV400 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Product Content Correction
Product Title
Draeger Safety X-Pect Glasses >
Spot an opportunity for improvement?Share a Content Correction