Learn More
Lovibond™ DPD No. 3 Evo Tablets
This tablet is used in combination with the DPD No. 1 Tablet to measure Free and Total Chlorine concentrations in a water sample.
Brand:  Lovibond™ 511420BT
 View more versions of this product
Product Code. 17813971
  • 289.00 SEK / Pack of 100
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Description
  • Shelf-stable, analytical solution for testing water quality parameters with tablet reagents
  • Each tablet is protected from humidity, heat and other environmental interferences
  • Pharmaceutical-grade aluminum blister packaging individually seals up to 10 tablets on an individual strip
  • Easy to dispense blister packaging provides safe handling of the reagents
  • Accurate and precise dosing
  • Long shelf life
Specifications
Specifications
Chlorine
Water Quality Analysis
100 Tests
DPD
Tablet
Lovibond™ Colorimeters and Spectrophotometers
10 mL
2 min