missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC1 Monoclonal Antibody (144), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512071
This item is not returnable.
View return policy
Description
A suggested positive control for this product is HT29, LS174T, HeLa cells or normal testis.
XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.
Specifications
XRCC1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P18887 | |
XRCC1 | |
Recombinant human XRCC1 protein | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
144 | |
Unconjugated | |
XRCC1 | |
DNA repair protein XRCC1; RCC; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross complementing 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1; XRCC1; Xrcc-1 | |
Mouse | |
Protein A | |
RUO | |
7515 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
XRCC1 Monoclonal Antibody (144), Invitrogen™ > 500 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction