Learn More
Xylene Cyanol FF 85%, Thermo Scientific Chemicals
Product Code. 10414511
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
500 g
Packaging:
Glass bottle
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4463-44-9
Molecular Formula C25H28N2NaO7S2
Molecular Weight (g/mol) 555.616
MDL Number MFCD00019481
InChI Key JKPDSWLBCLGXEQ-UHFFFAOYSA-N
Synonym C.I. 43535, Cyanol FF
PubChem CID 131854036
IUPAC Name 4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonic acid;sodium
SMILES CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)O)O)C.[Na]
Specifications
Dye Content 80% min.
Infrared Spectrum Authentic
Formula Weight 554.61
Grade Extra Pure
Packaging Glass bottle
Color Blue to Red
Quantity 500 g
Physical Form Crystalline Powder
Chemical Name or Material Xylene cyanol, 85%
compliance-icons
Product Identifier
  • Xylene cyanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.