missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, 99+%, Thermo Scientific Chemicals

346.73 SEK - 8614.98 SEK

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10055663
View Documents Promotion Details
Thermo Scientific Acros
225980250
25 g Plastic bottle
346.73 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
10276610
View Documents
Thermo Scientific Acros
225981000
100 g Plastic bottle
892.27 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
10371514
View Documents
Thermo Scientific Acros
225985000
500 g Plastic bottle
2414.09 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
10743455
View Documents
Thermo Scientific Acros
225980025
2.5 kg Plastic bottle
8614.98 SEK
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

87-99-0
152.15
HEBKCHPVOIAQTA-UHFFFAOYNA-N
6912
OCC(O)C(O)C(O)CO
C5H12O5
MFCD00064291,MFCD00064292
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
(2S,4R)-pentane-1,2,3,4,5-pentol

Specifications

87-99-0
White
Authentic
C5H12O5
MFCD00064291,MFCD00064292
15, 10283
Solubility in water: 1700g/L (20°C). Other solubilities: soluble in pyridine,less soluble in alcohols
OCC(O)C(O)C(O)CO
152.15
152.15
Crystalline Powder
92.0°C to 96.0°C
215.0°C to 217.0°C
2% max. Other polyols (GC)
HOCH2[CH(OH)]3CH2OH
01, 531
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
HEBKCHPVOIAQTA-UHFFFAOYNA-N
(2S,4R)-pentane-1,2,3,4,5-pentol
6912
99+%
Xylitol, 99+%
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

missing translation for 'einecsNumber' : 201-788-0

missing translation for 'rtecsNumber' : ZF0800000

missing translation for 'tsca' : TSCA

Product Identifier
  • Xylitol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only