missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAP70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ZAP70 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894824
|
Proteintech
15592-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884824
|
Proteintech
15592-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Interacts with phosphorylated C-terminal heptapeptide repeat domain (CTD) of the largest RNA polymerase II subunit POLR2A, and participates in dephosphorylation of the CTD. Transcriptional regulator which enhances expression of CCND1. Promotes binding of RNA polymerase II to the CCDN1 promoter and to the termination region before the poly-A site but decreases its binding after the poly-A site. Prevents RNA polymerase II from reading through the 3′ end termination site and may allow it to be recruited back to the promoter through promotion of the formation of a chromatin loop. Also enhances the transcription of a number of other cell cycle-related genes including CDK2, CDK4, CDK6 and cyclin-E but not CDKN1A, CDKN1B or cyclin-A. Promotes cell proliferation.Specifications
ZAP70 | |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P43403, P43404 | |
22637, 301348, 7535 | |
ZAP70 Fusion Protein Ag7943 | |
Primary | |
-20°C | |
ZAP70 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
70 kDa zeta associated protein, SRK, STD, Syk related tyrosine kinase, Tyrosine protein kinase ZAP 70, TZK, ZAP 70, ZAP70 | |
ZAP70 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |