missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZG16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17397-1-AP-20UL
This item is not returnable.
View return policy
Specifications
ZG16 | |
Polyclonal | |
Unconjugated | |
ZG16 | |
hZG16, JCLN, JCLN1, Secretory lectin ZG16, ZG16, ZG16A, Zymogen granule protein 16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171449, 653808, 69036 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60844, Q8CJD3, Q8K0C5 | |
Zg16 | |
ZG16 Fusion Protein Ag11332 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction