missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc-alpha-2-glycoprotein Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13399-1-AP-20UL
This item is not returnable.
View return policy
Description
AZGP1 stimulates lipid degradation in adipocytes and causes the extensive fat losses associated with some advanced cancers. It may bind polyunsaturated fatty acids.Specifications
Zinc-alpha-2-glycoprotein | |
Polyclonal | |
Unconjugated | |
AZGP1 | |
AZGP1, ZA2G, ZAG, Zinc-alpha-2-glycoprotein, Zn alpha 2 glycoprotein, Zn alpha 2 GP, ZNGP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
563 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P25311 | |
AZGP1 | |
Zinc-alpha-2-glycoprotein Fusion Protein Ag4232 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zinc-alpha-2-glycoprotein Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction