missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Carbonate, MP Biomedicals
636.00 SEK - 1160.00 SEK
Chemical Identifiers
CAS | 3486-35-9 |
---|---|
Molecular Formula | CO3Zn |
Molecular Weight (g/mol) | 125.388 |
InChI Key | FMRLDPWIRHBCCC-UHFFFAOYSA-L |
Synonym | zinc carbonate, zincspar, carbonic acid, zinc salt 1:1, zinc monocarbonate, natural smithsonite, zinc carbonate 1:1, unii-k8290ptq4f, hydrozincite?, zinc carbonic acid, zinc 2+ ion carbonate |
PubChem CID | 19005 |
IUPAC Name | zinc;carbonate |
SMILES | C(=O)([O-])[O-].[Zn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11447591
|
MP Biomedicals
0210473780 |
100 g |
636.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457591
|
MP Biomedicals
0210473790 |
500 g |
1160.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
3486-35-9 | |
125.388 | |
zinc carbonate, zincspar, carbonic acid, zinc salt 1:1, zinc monocarbonate, natural smithsonite, zinc carbonate 1:1, unii-k8290ptq4f, hydrozincite?, zinc carbonic acid, zinc 2+ ion carbonate | |
zinc;carbonate |
CO3Zn | |
FMRLDPWIRHBCCC-UHFFFAOYSA-L | |
19005 | |
C(=O)([O-])[O-].[Zn+2] |
Specifications
3486-35-9 | |
zinc carbonate, zincspar, carbonic acid, zinc salt 1:1, zinc monocarbonate, natural smithsonite, zinc carbonate 1:1, unii-k8290ptq4f, hydrozincite?, zinc carbonic acid, zinc 2+ ion carbonate | |
C(=O)([O-])[O-].[Zn+2] | |
125.388 | |
125.4 |
CO3Zn | |
FMRLDPWIRHBCCC-UHFFFAOYSA-L | |
zinc;carbonate | |
19005 | |
Zinc Carbonate |
Safety and Handling
Recommended Storage : Room Temperature
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title