missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chromate, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
CAS | 13530-65-9 |
Quantity | 100 g |
Molecular Formula | CrO4Zn |
MDL Number | MFCD00036281 |
Synonym | zinc chromate, zinc chrome, zinc chromate c, zinc chromate o, zinc chromate t, zinc chromate z, zinc chromate am, zincro zto, basic zinc chromate, zinc chrome yellow |
InChI Key | NDKWCCLKSWNDBG-UHFFFAOYSA-N |
SMILES | [Zn++].[O-][Cr]([O-])(=O)=O |
IUPAC Name | zinc(2+) dioxochromiumbis(olate) |
Molecular Weight (g/mol) | 181.37 |
PubChem CID | 26089 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'