Learn More
Zinc phosphate hydrate, tech., Thermo Scientific Chemicals
955.00 SEK - 2775.00 SEK
Chemical Identifiers
CAS | 34807-26-6 |
---|---|
Molecular Formula | O8P2Zn3 |
Molecular Weight (g/mol) | 386.08 |
MDL Number | MFCD00036282 |
InChI Key | LRXTYHSAJDENHV-UHFFFAOYSA-H |
Synonym | zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc |
PubChem CID | 24519 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307327
|
Thermo Scientific Alfa Aesar
011589.36 |
500 g |
955.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317327
|
Thermo Scientific Alfa Aesar
011589.A3 |
2 kg |
2775.00 SEK
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zinc phosphate hydrate is used in dentistry. It is commonly used for luting permanent metal restorations and as a base for dental restorations. Zinc phosphate cement is also used for cementation of inlays, crowns, bridges, and orthodontic appliances and occasionally as a temporary restoration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34807-26-6 | |
386.08 | |
LRXTYHSAJDENHV-UHFFFAOYSA-H | |
24519 |
O8P2Zn3 | |
MFCD00036282 | |
zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc |
Specifications
34807-26-6 | |
O8P2Zn3 | |
zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc | |
LRXTYHSAJDENHV-UHFFFAOYSA-H | |
trizinc(2+) diphosphate | |
24519 | |
Technical |
Powder | |
MFCD00036282 | |
Soluble in acetic acid,dilute mineral acids,ammonia. Insoluble in water | |
[Zn++].[Zn++].[Zn++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
386.08 | |
386.05 (Anhydrous) | |
Zinc phosphate hydrate |
Safety and Handling
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 231-944-3
missing translation for 'rtecsNumber' : TD0590000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.