missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Phosphide, MP Biomedicals
Click to view available options
Quantity:
10 g
Specifications
Specifications
CAS | 1314-84-7 |
Quantity | 10 g |
Formula Weight | 258.1 |
Chemical Name or Material | Zinc Phosphide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'