Learn More
Emma Safety Footwear Zion Safety Shoes
Chemical resistant with highly visible 3M™ reflective strip. The impenetrable sole protects feet from sharp and metal objects while the steel toe cap and reinforced polyurethane toe cover offer protection from falling objects. Microfiber components stand up to cement and remains flexible.
1215.00 SEK
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Color | Black |
Design | Zion |
Liner Material | Hydro-Tec™ Sanitized Silver |
Material | Microfiber |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15908291
|
Emma Safety Footwear
MM30084837 |
37 | N/A | ||||||
15918291
|
Emma Safety Footwear
MM30084838 |
38 | N/A | ||||||
15928291
|
Emma Safety Footwear
MM30084839 |
39 | N/A | ||||||
15938291
|
Emma Safety Footwear
MM30084840 |
40 | N/A | ||||||
15948291
|
Emma Safety Footwear
MM30084841 |
41 | N/A | ||||||
15958291
|
Emma Safety Footwear
MM30084842 |
42 | N/A | ||||||
15968291
|
Emma Safety Footwear
MM30084843 |
43 | N/A | ||||||
15978291
|
Emma Safety Footwear
MM30084844 |
44 | N/A | ||||||
15988291
|
Emma Safety Footwear
MM30084845 |
45 | N/A | ||||||
15998291
|
Emma Safety Footwear
MM30084846 |
46 | N/A | ||||||
15908301
|
Emma Safety Footwear
MM30084847 |
47 | N/A | ||||||
15918301
|
Emma Safety Footwear
MM30084848 |
48 | N/A | ||||||
15928301
|
Emma Safety Footwear
MM30084849 |
49 | N/A | ||||||
Description
- S3-Safety shoe with sole plate for wet work
- Microfiber construction
- GripForce™ Duo Heat Resistant Outsole
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
EN ISO 20345:2011 | |
Zion | |
Microfiber |
Black | |
Hydro-Tec™ Sanitized Silver |