missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF238 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1935.00 SEK - 4555.00 SEK
Specifications
Antigen | ZNF238 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18383225
|
Bio-Techne
NBP3-03476-20UL |
20 μL |
1935.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18323034
|
Bio-Techne
NBP3-03476-100UL |
100 μL |
4555.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF238 Polyclonal specifically detects ZNF238 in Human samples. It is validated for Western Blot.Specifications
ZNF238 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
10472 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
NIZP1, NSD1 (nuclear receptor binding SET-domain containing 1)-interacting zinc fingerprotein 1, zinc finger protein 496, Zinc finger protein with KRAB and SCAN domains 17, ZKSCAN17MGC15548 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 140-360 of human ZNF238 (NP_006343.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title