missing translation for 'onlineSavingsMsg'
Learn More

ZNF549 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

1935.00 SEK - 4555.00 SEK

Specifications

Antigen ZNF549
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18391897
View Documents
Bio-Techne
NBP3-04628-20UL
20 μL
1935.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18332024
View Documents
Bio-Techne
NBP3-04628-100UL
100 μL
4555.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF549 Polyclonal specifically detects ZNF549 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ZNF549
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
256051
IgG
Affinity purified
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Human
FLJ34917, zinc finger protein 549
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ZNF549 (NP_001186224.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.