missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF552 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1925.00 SEK - 4460.00 SEK
Specifications
Antigen | ZNF552 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18323214
|
Bio-Techne
NBP3-09925-25UL |
25 μg |
1925.00 SEK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18399955
|
Bio-Techne
NBP3-09925-100UL |
100 μg |
4460.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF552 Polyclonal specifically detects ZNF552 in Human samples. It is validated for Western Blot.Specifications
ZNF552 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
ZNF552 zinc finger protein 552 | |
The immunogen is a synthetic peptide directed towards the middle region of Human ZNF552 (NP_079038). Peptide sequence RSGLLQQEATHTGKSNSKTECVSLFHGGKSHYSCGGCMKHFSTKDILSQH | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
79818 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title