accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1000994-95-5

CAS RN 1000994-95-5

IUPAC Name: 1-bromo-4-(1,1-difluoroethyl)benzene
Synonyms: 1-bromo-4-(1,1-difluoroethyl)benzene
Molecular Weight (g/mol): 221.05
Molecular Formula: C8H7BrF2
InChi Key: QXIBKCFAFRHORF-UHFFFAOYSA-N
SMILES: CC(F)(F)C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products