accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1001264-89-6

CAS RN 1001264-89-6

IUPAC Name: 2-(4-chlorophenyl)-1-(4-{7-hydroxy-5-methyl-5H,6H,7H-cyclopenta[d]pyrimidin-4-yl}piperazin-1-yl)-3-[(propan-2-yl)amino]propan-1-one
Synonyms: 2-(4-chlorophenyl)-1-(4-{7-hydroxy-5-methyl-5H,6H,7H-cyclopenta[d]pyrimidin-4-yl}piperazin-1-yl)-3-(isopropylamino)propan-1-one
Molecular Weight (g/mol): 458
Molecular Formula: C24H32ClN5O2
InChi Key: GRZXWCHAXNAUHY-UHFFFAOYNA-N
SMILES: CC(C)NCC(C(=O)N1CCN(CC1)C1=NC=NC2=C1C(C)CC2O)C1=CC=C(Cl)C=C1
Color 
  • (7)
Grade 
  • (7)

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (7)
  • (7)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products