accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1020056-98-7

CAS RN 1020056-98-7

IUPAC Name: 2,3-dibromopyridine-4-carboxylic acid
Synonyms: 2,3-dibromopyridine-4-carboxylic acid
Molecular Weight (g/mol): 280.9
Molecular Formula: C6H3Br2NO2
InChi Key: ICISUZSRQHUJKX-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)C(Br)=NC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products