accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 102029-74-3

CAS RN 102029-74-3

IUPAC Name: 2-{[1-(2-{2-[2-({1-[2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidin-2-yl}formamido)-3-phenylpropanamido]-3-methylbutanamido}-4-carboxybutanoyl)pyrrolidin-2-yl]formamido}-3-methylpentanoic acid
Synonyms: 2-{[1-(2-{2-[2-({1-[2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidin-2-yl}formamido)-3-phenylpropanamido]-3-methylbutanamido}-4-carboxybutanoyl)pyrrolidin-2-yl]formamido}-3-methylpentanoic acid
Molecular Weight (g/mol): 864.01
Molecular Formula: C44H61N7O11
InChi Key: ADBHAJDGVKLXHK-UHFFFAOYNA-N
SMILES: CCC(C)C(NC(=O)C1CCCN1C(=O)C(CCC(O)=O)NC(=O)C(NC(=O)C(CC1=CC=CC=C1)NC(=O)C1CCCN1C(=O)C(N)CC1=CC=C(O)C=C1)C(C)C)C(O)=O

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products