accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1034449-15-4

CAS RN 1034449-15-4

IUPAC Name: 1,3-dicyclohexyl-3H-1λ⁵,3-benzodiazol-1-ylium chloride
Synonyms: 1,3-dicyclohexyl-1λ⁵,3-benzodiazol-1-ylium chloride
Molecular Weight (g/mol): 318.89
Molecular Formula: C19H27ClN2
InChi Key: JACUDTKAZHCIPU-UHFFFAOYSA-M
SMILES: [Cl-].C1CCC(CC1)N1C=[N+](C2CCCCC2)C2=CC=CC=C12

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products