accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1051487-82-1

CAS RN 1051487-82-1

IUPAC Name: N'-[1-(3-bromophenyl)ethylidene]-2-oxo-4-phenylpyrrolidine-3-carbohydrazide
Synonyms: N'-[1-(3-bromophenyl)ethylidene]-2-oxo-4-phenylpyrrolidine-3-carbohydrazide
Molecular Weight (g/mol): 400.28
Molecular Formula: C19H18BrN3O2
InChi Key: RQKXQCSEZPQBNZ-UHFFFAOYNA-N
SMILES: CC(=NNC(=O)C1C(CNC1=O)C1=CC=CC=C1)C1=CC(Br)=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products