accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1073372-01-6

CAS RN 1073372-01-6

IUPAC Name: 1-{4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazin-1-yl}ethan-1-one
Synonyms: 1-{4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazin-1-yl}ethanone
Molecular Weight (g/mol): 331.22
Molecular Formula: C17H26BN3O3
InChi Key: RNFCDPMIZSSWDQ-UHFFFAOYSA-N
SMILES: CC(=O)N1CCN(CC1)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products