man

Test

CAS RN 1092460-88-2

FBrOFFF

CAS RN 1092460-88-2

IUPAC Name: 2-(bromomethyl)-4-fluoro-1-(trifluoromethoxy)benzene
Synonyms: 2-(bromomethyl)-4-fluoro-1-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 273.03
Molecular Formula: C8H5BrF4O
InChi Key: VUUGICQPCOWAIC-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=C(OC(F)(F)F)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results