man

Test

CAS RN 109944-15-2

HOOHOHOHHNOON

CAS RN 109944-15-2

IUPAC Name: (5R,6R,7S,8R,8aS)-6,7,8-trihydroxy-5-(hydroxymethyl)-octahydroimidazo[1,2-a]pyridine-2,3-dione
Synonyms: kifunensine
Molecular Weight (g/mol): 232.19
Molecular Formula: C8H12N2O6
InChi Key: OIURYJWYVIAOCW-PQMKYFCFSA-N
SMILES: OC[C@@H]1[C@@H](O)[C@H](O)[C@H](O)[C@H]2NC(=O)C(=O)N12

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results