CAS RN 111358-88-4
CAS RN 111358-88-4
IUPAC Name:
(15S,16S,18R)-16-hydroxy-16-(hydroxymethyl)-15-methyl-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8,10,12,20(25),21,23-nonaen-3-one
Synonyms:
lestaurtinib
Molecular Weight (g/mol):
439.47
Molecular Formula:
C26H21N3O4
InChi Key:
UIARLYUEJFELEN-LROUJFHJSA-N
SMILES:
C[C@]12O[C@H](C[C@]1(O)CO)N1C3=C(C=CC=C3)C3=C1C1=C(C4=C3C(=O)NC4)C3=CC=CC=C3N21