man

Test

CAS RN 12007-60-2

BBOOOOOBBOOLiLi

CAS RN 12007-60-2

IUPAC Name: tetraboron heptahydrate dilithium
Synonyms: lithium borate
Molecular Weight (g/mol): 183.23
Molecular Formula: B4H14Li2O7
InChi Key: DPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES: [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
Quantity 
  • (10)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (11)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (10)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (11)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)