CAS RN 1256355-32-4
CAS RN 1256355-32-4
IUPAC Name:
(4-amino-3-fluorophenyl)boronic acid hydrochloride
Synonyms:
4-amino-3-fluorophenylboronic acid hydrochloride
Molecular Weight (g/mol):
191.39
Molecular Formula:
C6H8BClFNO2
InChi Key:
NXCNVCKJELIVDM-UHFFFAOYSA-N
SMILES:
Cl.NC1=C(F)C=C(C=C1)B(O)O
1
–
1
of
1
results
1
4-Amino-3-fluorophenylboronic acid hydrochloride, 97%, Thermo Scientific Chemicals
CAS: 1256355-32-4 Molecular Formula: C6H7BFNO2·ClH Molecular Weight (g/mol): 191.4 InChI Key: NXCNVCKJELIVDM-UHFFFAOYSA-N Synonym: 4-amino-3-fluorophenylboronic acid hydrochloride,4-amino-3-fluorophenylboronic acid, hydrochloride,4-amino-3-fluorophenyl boronic acid hydrochloride,4-amino-3-fluorophenylboronic acid,hydrochloride,4-amino-3-fluorophenylboronic acid,hcl,4-amino-3-fluorophenylboronic acid, hcl,4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1,boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1 PubChem CID: 46738020