man

Test

CAS RN 129-46-4

NaNaNaNaNaNaH3CNHOHNOHNONHCH3OHNSOOOSOOOSOOOONHSOOOSOOOSOOO

CAS RN 129-46-4

IUPAC Name: hexasodium 8-{4-methyl-3-[3-({[3-({2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl}carbamoyl)phenyl]carbamoyl}amino)benzamido]benzamido}naphthalene-1,3,5-trisulfonate
Synonyms: hexasodium 8-{4-methyl-3-[3-({[3-({2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl}carbamoyl)phenyl]carbamoyl}amino)benzamido]benzamido}naphthalene-1,3,5-trisulfonate
Molecular Weight (g/mol): 1429.15
Molecular Formula: C51H34N6Na6O23S6
InChi Key: VAPNKLKDKUDFHK-UHFFFAOYSA-H
SMILES: [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].CC1=CC=C(C=C1NC(=O)C1=CC(NC(=O)NC2=CC(=CC=C2)C(=O)NC2=CC(=CC=C2C)C(=O)NC2=C3C(C=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C2)S([O-])(=O)=O)=CC=C1)C(=O)NC1=C2C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)