man

Test

CAS RN 130209-90-4

CH3OOClNSHCl

CAS RN 130209-90-4

IUPAC Name: methyl 2-(2-chlorophenyl)-2-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}acetate hydrochloride
Synonyms: clopidogrel hydrochloride
Molecular Weight (g/mol): 358.28
Molecular Formula: C16H17Cl2NO2S
InChi Key: XIHVAFJSGWDBGA-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N1CCC2=C(C1)C=CS2)C1=CC=CC=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results