man

Test

CAS RN 1312100-75-6

ONBrSOOCl

CAS RN 1312100-75-6

IUPAC Name: 4-[(5-bromopyridin-2-yl)oxy]benzene-1-sulfonyl chloride
Synonyms: 4-[(5-bromopyridin-2-yl)oxy]benzenesulfonyl chloride
Molecular Weight (g/mol): 348.6
Molecular Formula: C11H7BrClNO3S
InChi Key: PRPZOWBZYLSCTG-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(OC2=NC=C(Br)C=C2)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results