man

Test

CAS RN 13127-88-3

O2H2H2H2H2H2H

CAS RN 13127-88-3

IUPAC Name: 1-[(²H)oxy](²H₅)benzene
Synonyms: 1-[(²H)oxy](²H₅)benzene
Molecular Weight (g/mol): 100.15
Molecular Formula: C6H6O
InChi Key: ISWSIDIOOBJBQZ-QNKSCLMFSA-N
SMILES: [2H]OC1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]
For Use With (Equipment) 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)