man

Test

CAS RN 1313-84-4

SNaNaH2OH2OH2OH2OH2OH2OH2OH2OH2O

CAS RN 1313-84-4

IUPAC Name: disodium nonahydrate sulfanediide
Synonyms: disodium nonahydrate sulfanediide
Molecular Weight (g/mol): 240.17
Molecular Formula: H18Na2O9S
InChi Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
Grade 
  • (4)
  • (8)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (10)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (4)
  • (8)
  • (1)
  • (1)
  • (2)
  • (10)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)