man

Test

CAS RN 13596-46-8

NH4NH4CoOSOOOOSOOO

CAS RN 13596-46-8

IUPAC Name: λ²-cobalt(2+) diammonium disulfate
Synonyms: λ²-cobalt(2+) diammonium disulfate
Molecular Weight (g/mol): 287.12
Molecular Formula: CoH8N2O8S2
InChi Key: NUHXWPRATKOXDB-UHFFFAOYSA-L
SMILES: [NH4+].[NH4+].[Co++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Quantity 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results