man

Test

CAS RN 142808-15-9

BrFFFF

CAS RN 142808-15-9

IUPAC Name: 4-bromo-2-fluoro-1-(trifluoromethyl)benzene
Synonyms: 4-bromo-2-fluoro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 243
Molecular Formula: C7H3BrF4
InChi Key: OEPBVXQEVBURGC-UHFFFAOYSA-N
SMILES: FC1=C(C=CC(Br)=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results